Product Image
Thermo Scientific™
Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)
Isolate and identify Neisseria gonorrhoeae and Neisseria meningitidis with this selective, ready-to-use solid medium
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R01882100/Pk.
R0188010/Pk.
Catalog number R01882
Price (USD)
-
Quantity:
100/Pk.
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel™ Thayer Martin Agar Modified (with nystatin, colistin, vancomycin, trimethoprim)
Thayer Martin Agar is a selective medium containing antibiotics to isolate Neisseria gonorrhoeae and Neisseria meningitidis, while suppressing commensal microbial flora present in the anal canal, vagina, pharynx and other sites1
Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim) is a solid medium recommended to isolate Neisseria gonorrhoeae and Neisseria meningitidis.
  • Isolates: Neisseria gonorrhoeae and Neisseria meningitidis
  • Selective: Colistin, vancomycin and trimethoprim suppress the growth of commensal microbial flora present in the anal canal, vagina, pharynx and other sites. Nystatin inhibits Candida albicans, colistin inhibits most gram-negative microorganisms, including Pseudomonas spp. and vancomycin inhibits gram-positive cocci.
  • Ready-to-use: Convenience of prepared media.
Thayer Martin Agar (Modified) contains hemoglobin, which provides the X factor (hemin). GCHI Enrichment provides the V factor, vitamins, amino acids, coenzymes, and dextrose. Selective agents, vancomycin and colistin, inhibit gram-positive cocci and gram-negative bacilli, respectively. Thayer Martin Agar (Modified) contains nystatin, which has proven effective in suppressing the growth of Candida albicans. Trimethoprim lactate suppresses the swarming of Proteus species. Dextrose enhances the growth of gonococci. The JEMBECTM system contains a tablet, which liberates carbon dioxide into the enclosed pack when in contact with water vapour present in the medium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
1. Thayer, J.D. and Martin, J.E. Jr. 1964. Public Health Rep. 79:49.
Specifications
DescriptionThayer Martin Agar Modified
Format12mm x 85mm Monoplate
Quantity100/Pk.
Product TypeAgar Plate
Unit SizeEach