dNTP Mix (10 mM each), 10 x 1 mL - FAQs
dNTP Mix (10 mM each), 10 x 1 mL - FAQs
View additional product information for dNTP Mix (10 mM each) - FAQs (R0191, R0194, R0192, R0193)
1 product FAQs found
What concentration of dNTPs is typically used in a PCR amplification?
The four dNTPs (dATP, dCTP, dTTP, dGTP) are used at a final concentration of 200 µM each or a total of 0.8 mM in standard amplifications. For PCR amplifications that require high fidelity, lower dNTP concentrations (on the order of 10-50 µM) work optimally.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.