Remel™ Biplates: Anaerobic Reducible through Levine Agars
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Have Questions?
Change view
Catalog Number
Quantity
Media Type
R02066
100/Pk.
Columbia CNA/MacConkey Agar
R02055
10/Pk.
Chocolate Agar/Chocolate Agar
R02058
10/Pk.
Chocolate /Thayer Martin Agar
R02065
10/Pk.
Columbia CNA/MacConkey Agar
4 Options
Catalog number R02066
Price (USD)
-
Quantity:
100/Pk.
Media Type:
Columbia CNA/MacConkey Agar
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel™ Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.