Remel™ Biplates, HE through V Agar, HE Agar, XLD Agar
Thermo Scientific™
Remel™ Biplates, HE through V Agar, HE Agar, XLD Agar
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Have Questions?
Catalog number R02086
Price (USD)
-
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
  • Ready-to-use <
Specifications
DescriptionHE Agar; XLD Agar
FormatBiplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach