Use for the determination of indole production and nitrate reduction by anaerobic bacteria.
Have Questions?
Catalog Number
Quantity
R05059
20 x 5 mL
Catalog number R05059
Price (USD)
-
Quantity:
20 x 5 mL
Thermo Scientific™ Remel PRAS Indole Nitrate Medium (VPI) (Pre-Reduced Anaerobic Sterilized) is for the determination of indole production and nitrate reduction by anaerobic bacteria.
Specifications
DescriptionP.R.A.S. Indole Nitrate Medium ; Format: ; pack of 20