Search
Search
| Catalog Number | Quantity | Format | Media Type |
|---|---|---|---|
| R05176 | 100 x 3 mL | 16mm x 125mm Tube | PRAS PY Bile |
| R05161 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Base Control |
| R05175 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Bile |
| R05191 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Glucose |
| R05194 | 20 x 10 mL | 15 x 103mm Tube | PRAS PY Glucose |
| R05205 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Inositol |
| R05171 | 20 x 3 mL | 16mm x 125mm Tube, Stopper | PRAS PY Arabinose |
| R05181 | 100 x 3 mL | 16mm x 125mm Tube | PRAS PY Esculin |
| R05192 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Glucose Agar |
| R05189 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Galactose |
| R05218 | 100/Pk. | 16mm x 125mm Tube | PRAS PY Maltose |
| R05211 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Lactate (VPI) |
| R05213 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Lactose (VPI) |
| R05201 | 20 x 3 mL | 16mm x 125mm Tube | PRAS PY Glycogen (VPI) |
Differentiate anaerobic bacteria using Thermo Scientific™ Remel PRAS (Pre-Reduced Anaerobic Sterilized) PY Media.