What is the stability of TurboFECT after being frozen?
TurboFect is a sterile solution of a proprietary cationic polymer in water. Its performance is not affected by a few freeze-thaw cycles. If TurboFECT arrives frozen or is accidentally stored at -20 degrees C, simply thaw and mix it thoroughly before use. For long-term storage, keep TurboFECT at 4 degrees C as recommended.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.