Remel™ Andrade's Broth
Thermo Scientific™

Remel™ Andrade's Broth

Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMedia Type
R060142Andrade Broth w/ 1% Maltose
R060134Andrade Broth w/ 1% Inulin
R060126Andrade Broth w/ 1% Glycerol
R060150Andrade Broth w/ 1% Mannose
Catalog number R060142
Price (USD)
-
Media Type:
Andrade Broth w/ 1% Maltose

Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.

Specifications
Format15mm x 103mm Tube
Media TypeAndrade Broth w/ 1% Maltose
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach