Remel™ BHI Broth w/Fildes Enrichment
Thermo Scientific™

Remel™ BHI Broth w/Fildes Enrichment

Thermo Scientific™ Remel BHI Broth w/Fildes Enrichment (Brain Heart Infusion Broth) is for the cultivation of Haemophilus spp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R06028220 x 5 mL
R060280100 x 5 mL
Catalog number R060282
Price (USD)
-
Quantity:
20 x 5 mL

Thermo Scientific™ Remel BHI Broth w/Fildes Enrichment (Brain Heart Infusion Broth) is for the cultivation of Haemophilus spp.

Specifications
DescriptionBHI Broth w/Fildes Enrichment
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeBroth Tube
Unit SizeEach