Remel™ GN Broth (Hajna)
Thermo Scientific™

Remel™ GN Broth (Hajna)

Contains selective enrichment for isolation of gram-negative bacilli, especially Salmonella and Shigella.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R06099220 x 5 mL
R0838220 x 10 mL
R060990100 x 5 mL
R08380100 x 10 mL
Catalog number R060992
Price (USD)
-
Quantity:
20 x 5 mL

Thermo Scientific™ Remel GN Broth (Hajna) contains selective enrichment for isolation of gram-negative bacilli, especially Salmonella and Shigella.

Specifications
Certifications/ComplianceIndustrial Reference: USDA.
DescriptionGN Broth
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach