Product Image
Thermo Scientific™

Remel™ Heart Infusion Broth with BCP, 1% Cellobiose

Use for the determination of fermentation reactions of microorganisms, especially streptococci and enterococci.
Have Questions?
Catalog NumberQuantity
R06106420 x 5 mL
Catalog number R061064
Price (USD)
-
Quantity:
20 x 5 mL

Thermo Scientific™ Remel Heart Infusion Broth with BCP, 1% Cellobiose is used for determination of fermentation reactions of microorganisms, especially streptococci and enterococci.

Specifications
DescriptionHeart Infusion Broth w/BCP, 1% Cellobiose
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach