DNA Gel Loading Dye (6X) - FAQs
DNA Gel Loading Dye (6X) - FAQs
View additional product information for DNA Gel Loading Dye (6X) - FAQs (R0611)
2 product FAQs found
How much of the 6X Loading Dye should I add to my DNA sample?
Add 1/6 volume of 6X DNA Loading Dye to your DNA sample.
What molecular weight does bromophenol blue and xylene cyanol FF migrate in the 6X DNA loading dye (Cat. No. R0611)?
In a 1% agarose gel, bromophenol blue comigrates with approximately 300 bp fragment and xylene cyanol FF comigrates with approximately 4000 bp fragment.