Product Image
Thermo Scientific™

Heart Infusion Broth w/BCP, 1% Melibiose (Brom Cresol Purple), D - 5mL

Thermo Scientific™ Remel Heart Infusion Broth with BCP, 1% Melibiose (Brom Cresol Purple) for determination of fermentation reactions of microorganisms, especially streptococci and enterococci.
Have Questions?
Catalog NumberQuantity
R06111220 x 5 mL
Catalog number R061112
Price (USD)
-
Quantity:
20 x 5 mL
Thermo Scientific™ Remel Heart Infusion Broth with BCP, 1% Melibiose (Brom Cresol Purple) for determination of fermentation reactions of microorganisms, especially streptococci and enterococci.
Specifications
DescriptionHeart Infusion Broth w/BCP
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach