Search
Search

Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
| Catalog Number | Quantity | Format |
|---|---|---|
| R061478 | 100 x 4 mL | 15mm x 103mm Tube |
| R061470 | 100 x 1 mL | 15mm x 103mm Tube |
| R061472 | 20 x 1 mL | 15mm x 103mm Tube |
| R061480 | 20 x 4 mL | 15mm x 103mm Tube |
| R112474 | 4 x 500 mL | Borosilicate Bottle |
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. MH Broth was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3.
Cultivate a wide variety of microorganisms using Mueller Hinton Broth.
Mueller Hinton Broth is prepared with beef extract and acid digest of casein that supply amino acids, nitrogenous substances, vitamins, carbon, sulphur and other nutrients which are necessary for bacterial growth. Starch serves as a growth factor and a protective colloid that neutralizes toxic products which may be formed during the growth of bacteria.