Product Image
Thermo Scientific™
Remel™ Nitrate Agar
Thermo Scientific™ Remel Nitrate Agar is for the determination of nitrate reduction by bacteria.
Have Questions?
Catalog NumberQuantity
R06152620/Pk.
Catalog number R061526
Price (USD)
-
Quantity:
20/Pk.
Thermo Scientific™ Remel Nitrate Agar is for the determination of nitrate reduction by bacteria.
Specifications
DescriptionNitrate Agar
Format15mm x 103mm Tube - Slant
Quantity20/Pk.
Product TypeAgar
Unit SizeEach