Thermo Scientific™ Remel OF King Medium with 1% Galactose is used for determination of oxidative and fermentative metabolism of carbohydrate by gram-negative bacilli.
Have Questions?
Catalog Number
Quantity
R061930
4 x 20 mL
Catalog number R061930
Price (USD)
-
Quantity:
4 x 20 mL
Thermo Scientific™ Remel OF King Medium with 1% Galactose is for the determination of oxidative and fermentative metabolism of carbohydrates by gram-negative bacilli.