Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Thermo Scientific™ Remel Phenol Red Agar Base Control is a control for phenol red agars.
Description | Phenol Red Agar Base Control |
Format | 15mm x 103mm Tube - Slant |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Format | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R062502 | Each | 15mm x 103mm Tube - Slant | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Thermo Scientific™ Remel Phenol Red Agar Base Control is a control for phenol red agars.