Product Image
Thermo Scientific™
Remel™ Sellers Agar
Provides differentiation of nonfermentative gram-negative bacilli based on fluorescence, oxidation of dextrose, production of nitrogen gas and arginine utilization.
Have Questions?
Catalog NumberFormat
R06452215mm x 103mm Tube - Slant
Catalog number R064522
Price (USD)
-
Format:
15mm x 103mm Tube - Slant
Thermo Scientific™ Remel Sellers Agar provides differentiation of nonfermentative gram-negative bacilli based on fluorescence, oxidation of dextrose, production of nitrogen gas and arginine utilization.
Specifications
DescriptionSellers Agar
Format15mm x 103mm Tube - Slant
Quantity20/Pk.
Product TypeAgar
Unit SizeEach