Product Image
Thermo Scientific™

Thioglycollate Gelatin Medium

Identify microorganisms, especially anaerobes, based on the ability to liquefy gelatin with ready-to-use Thioglycollate Gelatin Medium.

Have Questions?
Catalog NumberQuantity
R06466820 x 5 mL
Catalog number R064668
Price (USD)
-
Quantity:
20 x 5 mL

Identify microorganisms based on the ability to liquefy gelatin with Thermo Scientific™ Remel™ Thioglycollate Gelatin Medium. First described by Brewer for the cultivation of aerobic organisms and obligate anaerobes1,2, Brewer's formulation was modified by replacement of meat infusion with peptones to enhance growth.3,4. Gelatin is added to Thioglycollate Gelatin Medium for detection of gelatin liquefaction by anaerobes, microaerophiles and aerobic microorganisms. Most microorganisms grow luxuriantly in this medium regardless of their oxygen requirements.

Thioglycollate Gelatin Medium is suitable for cultivation of both aerobic and anaerobic organisms.

Sodium thioglycollate acts as a reducing agent which maintains low oxygen tension in the medium. A small amount of agar provides for anaerobic conditions in the depths of the tubes by convection current restriction. Gelatin serves as the substrate that allows for detection of gelatinase produced by certain organisms.

  • Ready-to-use: Convenience of prepared media.
  • Easy-to-interpret: If the medium remains liquid after refrigeration at 2-8 °C, it indicates a positive test. The medium remains solid in the case of a negative test.

Thioglycollate Gelatin Medium contains peptones and cysteine that supply nitrogen, carbon and trace elements necessary for bacterial growth. Dextrose provides a ready source of energy. Sodium thioglycolate acts as a reducing agent. Sodium chloride serves as the source of essential electrolytes and maintains osmotic equilibrium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Brewer, J.H. 1940. J. Am. Med Assoc. 115:598-600.
  2. Brewer, J.H. 1940. J. Bacteriol. 39:10.
  3. Brewer, J.H. 1943, J. Bacteriol. 46:395-397.
  4. Vera, H.D. 1944. J. Bacteriol. 47:59-68.
Specifications
DescriptionThioglycollate Gelatin Medium
Format15mm x 90mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach