Todd Hewitt Broth
Thermo Scientific™

Todd Hewitt Broth

Reduce media prep time while cultivating beta hemolytic streptococci and encouraging the production of antigenic streptococci hemolysins with pre-filled, ready-to-use tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatFill Volume
R06480520/Pk.15mm x 103mm Tube5 mL
R064804100/Pk.Tube5 mL
Catalog number R064805
Price (USD)
-
Quantity:
20/Pk.
Format:
15mm x 103mm Tube
Fill Volume:
5 mL
Cultivate beta hemolytic streptococci, encourage the production of antigenic streptococci hemolysins, and isolate pathogenic microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Todd Hewitt Broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth, available in multiple formats and formulations.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionTodd Hewitt Broth
Fill Volume5 mL
Format15mm x 103mm Tube
Quantity20/Pk.
Manufacturing OriginUnited States
Product TypeMicrobiology Media
Unit SizeEach