This product will no longer be available to order from April 1, 2025 and may currently be subject to a minimum order quantity and extended delivery time. Alternative product R064844 is available. Click the link to find out more.
Remel™ Todd Hewitt Broth w/Gentamicin, Nalidixic Acid, Blood
Thermo Scientific™
Remel™ Todd Hewitt Broth w/Gentamicin, Nalidixic Acid, Blood
Provides selective isolation of group B streptococci from clinical specimens containing mixed flora.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R06484620 x 5 mL
R064844100 x 5 mL
Catalog number R064846
Price (USD)
-
Quantity:
20 x 5 mL
Todd Hewitt Broth with Gentamicin, Nalidixic Acid and Blood (Trans-Vag Broth w/5% Sheep Blood) is used for selective isolation of group B streptococci from clinical specimens containing mixed flora.
  • Gentamicin and nalidixic acid inhibit some gram-negative bacteria
Specifications
DescriptionTodd Hewitt Broth; Trans-Vag Broth w/Blood;
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach