Search
Search

| Catalog Number | Quantity | Format | Packaging Type |
|---|---|---|---|
| R064860 | 100/Pk. | 15mm x 103mm Tube | 100/Pk. |
| R01917 | 100/Pk. | Monoplate | 100/Pk. |
| R01920 | 10/Pk. | Monoplate | 10/Pk. |
| R04080 | 10/Pk. | 15mm x 150mm Plate | 10/Pk. |
| R064862 | 20/Pk. | 15mm x 103mm Tube | 20/Pk. |
| R08930 | 100/Pk. | 20mm x 113mm Tube | 100/Pk. |
| R08932 | 20/Pk. | 20mm x 113mm Tube | 20/Pk. |
| R08934 | 100 x 10 mL | 20mm x 113mm Pour Tube | 100/Pk. |
| R09938 | 100 x 22 mL | 20mm x 150mm Pour Tube | 100/Pk. |
| R09940 | 20 x 22 mL | 20mm x 150mm Pour Tube | 20/Pk. |
| R110092 | 100/Pk. | 15mm x 100mm Plate | 100/Pk. |
| R112700 | 10 x 500 mL | BB - 500mL | 10/Pk. |
| R112703 | 10 x 100 mL | BRB - 100mL | 10/Pk. |
| R01922 | 10/Pk. | Monoplate Deep Pour | 10/Pk. |
Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.