Urea Broth For AFB
Thermo Scientific™

Urea Broth For AFB

Differentiate Mycobacterium species based on urea hydrolysis using ready-to-use liquid medium Urea Broth For AFB.

Have Questions?
Catalog NumberQuantity
R06522220 x 1.5 mL
Catalog number R065222
Price (USD)
-
Quantity:
20 x 1.5 mL

Differentiate Mycobacterium species based on urea hydrolysis with Thermo Scientific™ Remel™ Urea Broth For AFB. In 1979, Steadham developed the urease test to help in identification of mycobacteria1. This broth test helped to achieve more consistent results for urease activity. The determination of urea hydrolysis (release of ammonia) helps in the characterization of mycobacterial strains, in particular, to distinguish scotochromogens and nonphotochromogens.2-5

Urea Broth For AFB is a liquid medium used to detect Mycobacterium species based on urea hydrolysis. This is a highly buffered medium which helps in differentiation of mycobacterial strains after 7 days incubation.

Urea Broth For AFB contains peptone and dextrose that supply essential nutrients required for growth of mycobacteria. Urea acts as the substrate for hydrolysis. Polysorbate 80 is a surfactant which aids in the dispersion of aggregates. The low pH of 5.8 facilitates a clear-cut differentiation of positive and negative results. Phenol red (1%) serves as the pH indicator.

When urea is hydrolyzed ammonia is released and the resulting alkalinity is evidenced by the phenol red indicator. A dark pink or red color development is observed in the case of a positive test whereas broth remains yellow in the case of a negative test.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Steadham, J.E. 1979. J. Clin. Microbiol. 10:134-137.
  2. Toda, T. Y. Hagihara, and K. Takeya. 1960. Am. Rev. Respir. Dis. 83:775-761.
  3. Urabe, K. and H. Saito. 1964. Am. Rev. Respir. Dis. 90:266-267.
  4. Kent, P.T. and G.P. Kubica. 1985. Public Health Mycobacteriology-A Guide for the Level III Laboratory. U.S. Dept. of H.H.S. and CDC, Atlanta, GA.
  5. Isenberg, H.D. 2004. Clinical Microbiology Procedures Handbook. 2nd, Vol 2., Press, Washington, D.C.
Specifications
DescriptionUrea Broth For AFB
Format15mm x 103mm Tube
Quantity20 x 1.5 mL
Product TypeMicrobiology Media
Unit SizeEach