Search
Search

Cultivate aerobes and anaerobes using Thioglycollate Medium w/Dextrose, Hemin, Vitamin K.
| Catalog Number | Quantity |
|---|---|
| R07180 | 100 x 10 mL |
| R064732 | 20 x 5 mL |
| R064722 | 20 x 9 mL |
| R07182 | 20 x 10 mL |
| R064730 | 100 x 5 mL |
| R064720 | 100 x 9 mL |
Cultivate aerobes and anaerobes using Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose, Hemin, Vitamin K. This general-purpose medium was first described by Brewer in 19401, and consists of a small amount of agar and a reducing substance to initiate the growth of anaerobes.
Thioglycollate Medium w/Dextrose is a liquid medium recommended for use as a general purpose media. Sodium thioglycollate is the reducing agent which removes molecular oxygen from the medium and prevents the accumulation of peroxides which may be lethal to some organisms. Dextrose is added to the medium to study specific fermentation reactions.
Hemin is added as a source of X factor which stimulates the growth of many fastidious organisms and Vitamin K promotes growth of some Bacteroides species and gram-positive sporeformers.
This medium contains casein peptone and cystine that provide nitrogen and carbon essential for growth of aerobes and anaerobes. Dextrose is the source of fermentable carbohydrate. A small amount of agar is added to impede oxygen diffusion. Hemin stimulates the growth of fastidious organisms and Vitamin K promotes growth of some Bacteroides spp. and gram-positive sporeformers. Sodium thioglycolate is the reducing agent. Sulfahydryl groups inactivate mercury and other heavy metals and neutralize the antibacterial effect of some preservatives.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.