Remel™ Haemophilus Test Medium Broth (HTM Broth)
Product Image
Thermo Scientific™

Remel™ Haemophilus Test Medium Broth (HTM Broth)

Use for antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R0768120 x 10 mL
R112380500 mL
Catalog number R07681
Price (USD)
-
Request A Quote
Quantity:
20 x 10 mL

Thermo Scientific™ Remel Haemophilus Test Medium Broth (HTM Broth) is for the antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.

REF R112380 not intended for IVD use.
Specifications
DescriptionHaemophilus Test Medium Broth
Format16mm x 125mm Tube
Quantity20 x 10 mL
Product TypeMicrobiology Media
Unit SizeEach