Brain Heart Infusion (BHI) Agar
Thermo Scientific™

Brain Heart Infusion (BHI) Agar

Isolate and cultivate a wide variety of micro-organisms including pathogenic fungi this ready-to-use medium recommended by AOAC and APHA.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R0806220/Pk.20mm x 113mm Tube - Slant
R06023220/Pk.15mm x 103mm Tube
R08060100/Pk.20mm x 113mm Tube - Slant
R0906020/Pk.20mm x 150mm Pour Tube
Catalog number R08062
Price (USD)
-
Quantity:
20/Pk.
Format:
20mm x 113mm Tube - Slant

Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar. Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2

Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.

  • Recommended by AOAC and APHA.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

REF R112025 not intended for IVD use.
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF, EPA, IDF, SMD.
DescriptionBrain Heart Infusion Agar (BHI)
Format20mm x 113mm Tube - Slant
Quantity20/Pk.
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach