BHI Agar with Sheep Blood
Actual product may vary
Thermo Scientific™

BHI Agar with Sheep Blood

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with this nutritious medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R0806620/Pk.20mm x 113mm Tube
R0113610/Pk.Monoplate Deep Pour
R01139100/Pk.Monoplate Deep Pour
Catalog number R08066
Price (USD)
-
Quantity:
20/Pk.
Format:
20mm x 113mm Tube

Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with nutritious Thermo Scientific™ Remel™ BHI Agar with Sheep Blood. Due to the addition of Sheep blood, the BHI Agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media.

Use BHI Agar with Sheep Blood for the cultivation of a wide variety of microorganisms including pathogenic fungi.

  • Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
  • Ready-to-use: convenience of prepared media.

BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  

General References:

  1. Pasculle, A.W., J.C. Feeley, R.J. Gibson, L.G. Cordes, R.L. Myerowitz, C.M. Patton, G.W. Gorman, C.L. Carmack, J.W. Ezzell, and J.N. Dowling. 1980. J. Infect. Dis. 141:727-732. 5.
  2. Edelstein, P.H. 1981. J. Clin. Microbiol. 14:298-303.
  3. Versalovic, J., K.C. Carroll, G. Funke, J.H. Jorgensen, M.L. Landry, and D.W. Warnock. 2011. Manual of Clinical Microbiology. 10th ed. ASM Press, Washington, D.C.
  4. Microbiology Services. 2015. UK Standards for Microbiology Investigations. Bacteriology 9 (3): 1–27.
Specifications
DescriptionBHI Agar w/Sheep Blood
Format20mm x 113mm Tube
Quantity20/Pk.
Product TypeAgar Plate
Unit SizeEach