Product Image
Thermo Scientific™
Remel™ BHI Agar w/Chloramphenicol, Cycloheximide
Thermo Scientific™ Remel BHI Agar with Chloramphenicol and Cycloheximide is for the selective isolation of fungi and yeast.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R0806920/Pk.
R08068100/Pk.
Catalog number R08069
Price (USD)
-
Quantity:
20/Pk.
Thermo Scientific™ Remel BHI Agar with Chloramphenicol and Cycloheximide is for the selective isolation of fungi and yeast.
Chloramphenicol inhibits some gram-positive and gram-negative bacteria. Cycloheximide inhibits rapid growing saprophytic fungi.
Specifications
DescriptionBHI Agar w/Chloramphenicol, Cycloheximide
Format20mm x 113mm Tube - Slant
Quantity20/Pk.
Product TypeAgar Slants
Unit SizeEach