Product Image
Thermo Scientific™
Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R0807220/Pk.F - slant
R0114410/Pk.Monoplate Deep Pour
Catalog number R08072
Price (USD)
-
Quantity:
20/Pk.
Format:
F - slant
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
  • Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
  • Gentamicin inhibits some gram-negative bacteria
Specifications
DescriptionBHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
FormatF - slant
Quantity20/Pk.
Product TypeAgar
Unit SizeEach