Search
| Catalog Number | Format |
|---|---|
| R08073 | 20mm x 113mm Tube |
| R01142 | Monoplate Deep Pour |
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Chloramphenicol and Cycloheximide1. Isolate fastidious fungi and grow Actinomyces2 using BHI Agar with 5-10% sheep blood. Pathogenic fungi and Nocardia asteroides can be isolated using optimal concentrations of chloramphenicol3. Cycloheximide inhibits certain Aspergillus spp. and zygomycetes.
Use BHI Agar with Sheep Blood, Chloramphenicol and Cycloheximide1 for the cultivation of a wide variety of microorganisms, including pathogenic fungi.
BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose, which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References: