Search
Search

| Catalog Number | Quantity |
|---|---|
| R08382 | 20 x 10 mL |
| R060992 | 20 x 5 mL |
| R060990 | 100 x 5 mL |
| R08380 | 100 x 10 mL |
Thermo Scientific™ Remel GN Broth (Hajna) contains selective enrichment for isolation of gram-negative bacilli, especially Salmonella and Shigella.