Product Image
Thermo Scientific™
Remel™ Herrolds Medium
Thermo Scientific™ Remel Herrolds Medium is for the selective isolation of Mycobacterium paratuberculosis from cattle and other ruminants.
Have Questions?
Catalog NumberQuantity
R0838720/Pk.
Catalog number R08387
Price (USD)
-
Quantity:
20/Pk.
Thermo Scientific™ Remel Herrolds Medium is for the selective isolation of Mycobacterium paratuberculosis from cattle and other ruminants.
Not intended for IVD use.
Specifications
DescriptionHerrolds Medium
Format20mm x 113mm Tube - Slant
Quantity20/Pk.
Product TypeMicrobiology Media
Unit SizeEach