Product Image
Thermo Scientific™
Remel™ Littman Oxgall Agar w/Streptomycin
The Thermo Scientific™ Remel Littman Oxgall Agar with Streptomycin provides isolation of fungi, especially dermatophytes.
Have Questions?
Catalog NumberQuantity
R0846220/Pk.
Catalog number R08462
Price (USD)
-
Quantity:
20/Pk.
The Thermo Scientific™ Remel Littman Oxgall Agar with Streptomycin provides isolation of fungi, especially dermatophytes.
Specifications
DescriptionLittman Oxgall Agar w/Streptomycin
Format20mm x 113mm Tube - Slant
Quantity20/Pk.
Unit SizeEach