DTT (dithiothreitol), 25 g - FAQs
DTT (dithiothreitol), 25 g - FAQs
View additional product information for DTT (dithiothreitol) - FAQs (R0862, R0861)
1 product FAQs found
How do I prepare a 1 M DTT solution from powder and how should I store the solution?
Preparing 1 M DTT:
1) Dissolve 3.09 g DTT in 20 mL distilled H2O.
2) Dispense into 1 mL aliquots.
3) Store the aliquots at -20 degrees C.
Note: Upon thawing the vial, there may be some precipitation that can be re-dissolved by warming and mixing.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.