Product Image
Thermo Scientific™
Remel™ SABHI Agar with Blood and Gentamicin
The Thermo Scientific™ Remel SABHI Agar with Blood and Gentamicin provides selective isolation of pathogenic fungi.
Have Questions?
Catalog NumberQuantity
R0870320/Pk.
Catalog number R08703
Price (USD)
-
Quantity:
20/Pk.
Thermo Scientific™ Remel SABHI Agarwith Blood and Gentamicin provides selective isolation of pathogenic fungi.
Specifications
DescriptionSABHI Agar w/Blood, Gentamicin
Format20mm x 113mm Tube - Slant
Quantity20/Pk.
Unit SizeEach