Search
Search
Differentiate and identify Trichophyton species based on nutritional requirements with ready-to-use Trichophyton Agar No. 6.
| Catalog Number | Format | Media Type |
|---|---|---|
| R08921 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 6 |
| R09927 | G - PT | Trichophyton Agar No. 7 |
| R09923 | G - PT | Trichophyton Agar No. 6 |
| R09919 | G - PT | Trichophyton Agar No. 5 |
| R09915 | 20mm x 150mm Pour Tube | Trichophyton Agar No. 4 |
| R09911 | 20mm x 150mm Pour Tube | Trichophyton Agar No. 3 |
| R09907 | 20mm x 150mm Pour Tube | Trichophyton Agar No. 2 |
| R09903 | G - PT | Trichophyton Agar No. 1 |
| R08917 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 5 |
| R08913 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 4 |
| R08909 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 3 |
| R08905 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 2 |
| R08901 | 20mm x 113mm Tube - Slant | Trichophyton Agar No. 1 |
Differentiate and identify Trichophyton species based on nutritional requirements using Thermo Scientific™ Remel™ Trichophyton Agar No. 6 (Ammonium Nitrate Agar Base). Different Trichophyton species resemble one another closely in culture and are difficult to differentiate on the basis of colony formation or microscopic morphology alone. Georg and Camp developed Trichophyton Agars for differentiation and identification of Trichophyton isolates based on specific vitamin and amino acid requirements1.
Trichophyton Agars are to be used with Trichophyton isolates that have been presumptively identified by gross colony characteristics and microscopic morphology 2-5.
Trichophyton Agar No. 6 (Ammonium nitrate agar base) contains dextrose, monopotassium phosphate, magnesium sulfate and ammonium nitrate. Agar is the solidifying agent.
Trichophyton Agar No. 6 is the control medium used with Trichophyton Agar No. 7 to determine the histidine requirement.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.