Search
Search

Isolate and cultivate a wide variety of micro-organisms including pathogenic fungi this ready-to-use medium recommended by AOAC and APHA.
| Catalog Number | Quantity | Format |
|---|---|---|
| R09060 | 20/Pk. | 20mm x 150mm Pour Tube |
| R060232 | 20/Pk. | 15mm x 103mm Tube |
| R08060 | 100/Pk. | 20mm x 113mm Tube - Slant |
| R08062 | 20/Pk. | 20mm x 113mm Tube - Slant |
Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar. Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2
Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.