Sabouraud Dextrose Agar
Actual product may vary
Thermo Scientific™

Sabouraud Dextrose Agar

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatManufacturing Origin
R09744100/Pk.20 x 150mm Pour Tube (G-PT)United States
R01766100/Pk.Monoplate Deep PourUnited States
R0176810/Pk.Monoplate Deep PourUnited States
R09746100/Pk.20mm x 150mm Tube - SlantUnited States
R0974720/Pk.20mm x 150mm Tube - SlantUnited States
R11255110/Pk.BB - 500mLUnited States
R11256210/Pk.BRB - 100mLUnited States
Catalog number R09744
Price (USD)
-
Quantity:
100/Pk.
Format:
20 x 150mm Pour Tube (G-PT)
Manufacturing Origin:
United States

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.

Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF, USP.
ColorAmber
Fill Volume20 mL
Format20 x 150mm Pour Tube (G-PT)
Quantity100/Pk.
Shelf Life36-52 Weeks
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2 to 25°C