Search
| Catalog Number | Format | Media Type | Includes |
|---|---|---|---|
| R10055 | 1 oz. Bottle | Charcoal Blood Agar | Cephalexin |
| R01298 | Monoplate | Charcoal Blood Agar | Cephalexin |
Thermo Scientific™ Remel Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium) is used for selective isolation of Bordetella pertussis and Bordetella parapertussis.