Product Image
Thermo Scientific™
L-J Gruft
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R1010310/Pk.1 oz. Bottle
R08520100/Pk.20mm x 113mm Tube - Slant
R0852220/Pk.20mm x 113mm Tube - Slant
Catalog number R10103
Price (USD)
-
Quantity:
10/Pk.
Format:
1 oz. Bottle
Selectively isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel. Medium consisting of congo red and malachite green dyes was formulated by Lowenstein to cultivate mycobacteria1. This medium was modified by Jensen by replacing congo red with citrate and phosphate contents and increasing the malachite green concentration2. L-J Medium used for recovery of pathogenic Mycobacterium spp. was found to be superior to other media tested, including ATS (American Trudeau Society) Medium3,4.
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
  • Selective – Antibiotics in the medium make the medium selective for Mycobacterium while inhibiting contaminating bacteria.
  • Ready to use – Convenience of prepared media.
L-J Medium is a glycerated egg-potato medium. The glycerol and egg mixture provide fatty acids and protein required for the metabolism of mycobacteria. L-J Medium was modified by Gruft by adding ribonucleic acid as a growth stimulant and penicillin and nalidixic acid as selective inhibitors of contaminating bacteria5. Ribonucleic acid increases the percentage of positive cultures over the standard L-J Medium. The selectivity of L-J Medium was increased by incorporating cycloheximide, lincomycin, and nalidixic acid and was formulated by Petran and Vera6.
  1. Lowenstein, E. 1931. Zentralbl. Balteriol. Parasitenkd. Infektionskr. Hyg. Abt. 1 Orig. 120:127-129.
  2. Jensen, K.A. 1932. Zentralbl. Bakteroil. Infectionskr. Hyg. Abt. 1 Orig. 125:222-239.
  3. Cummings, M.M. 1951. J. Clin. 21:684-690.
  4. Hardy, A.V., F.P. Dunbar, M.B. Jeffries, J.O. Bond, and A.G. Lewis. 1958. Am. J. Public Health. 48:754-757.
Specifications
DescriptionLowenstein-Jensen Gruft. (LJ) w/penicillin, nalidixic acid
Format1 oz. Bottle
Quantity10/Pk.
Product TypePrepared Microbiology Tubes
Unit SizeEach