When using Calcium Crimson, AM, as an indicator of intracellular calcium flux, I am not getting a good degree of change. The cells also have GFP. What can I do?
This is a known drawback of Calcium Crimson, AM (as well as Calcium Orange, AM and Fura Red, AM, which are also in the same emission range). You can try increasing the concentration and washing out of any unlabeled dye from the media, to try to get better signal-to-background. If that fails, we recommend using Rhod-3 AM instead, which has a much better change in signal in that wavelength.