ReadyLabel™ Flex Antibody Labeling Kit, 5 x 20 μg of IgG - FAQs
ReadyLabel™ Flex Antibody Labeling Kit, 5 x 20 μg of IgG - FAQs
View additional product information for ReadyLabel™ Antibody Labeling Kits - FAQs (R10711, R10722, R10712, R10701, R10723, R10720, R10710, R10721, R10709, R10726, R10713, R10702, R10724, R10714)
1 product FAQs found
Do rat or bovine antibodies bind to the ReadyLabel columns?
ReadyLabel Columns bind poorly to rat and bovine antibodies, and are therefore, not recommended to use with the ReadyLabel Antibody Labeling Kits (Cat. Nos. R10701, R10702, R10709, R10710, R10711, R10712, R10713, R10714).