Brilliance™ CampyCount Agar
Thermo Scientific™

Brilliance™ CampyCount Agar

Accurately and efficiently identify and enumerate Campylobacter jejuni and C. coli from poultry samples on plate reader friendly, chromogenic plates.
Have Questions?
Catalog NumberFormatManufacturing Origin
R11016890mm MonoplateUnited States
Catalog number R110168
Price (USD)
-
Format:
90mm Monoplate
Manufacturing Origin:
United States
Accurately, specifically and easily enumerate Campylobacter jejuni and C. coli from poultry and related samples using Thermo Scientific™ Brilliance™ CampyCount Agar. The transparent medium, on which Campylobacter produce distinct dark red colonies, makes identification and enumeration significantly easier than on traditional charcoal or blood-containing agar. The transparency of the medium also allows enumeration using plate readers for even greater efficiency.

Campylobacter is endemic in many poultry populations, with Campylobacter jejuni and Campylobacter coli being responsible for the majority of infections. As legislation moves towards enumerative limits on Campylobacter in poultry, food producers will put pressure on poultry companies to provide meat with low numbers of Campylobacter to reduce human infection.

Brilliance CampyCount Agar has been validated and approved by MicroVal (according to ISO 16140: 2003 standard) against the reference method ISO/TS 10272-2: 2006 for the selective enumeration of thermotolerant Campylobacter species, in particular C. jejuni and C. coli, in poultry products. This study included both the Thermo Scientific O.B.I.S. Campy kit and Thermo Scientific™ Oxoid™ Dryspot Campylobacter test as alternative confirmation methods to those described in the reference method ISO/TS 10272-2: 2006.

  • Easy-to-read: chromogenic medium producing dark red colonies on a transparent background
  • Efficient and accurate: transparent medium allows enumeration on plate readers
  • Fast results: confirmed Campylobacter count in as little as 72 hours when used in conjunction with O.B.I.S. campy (ID0800) or Oxoid™ Campylobacter Test kit (DR0150)
  • Ready-to-use: convenience of prepared media

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery, we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Disclaimer: Not intended for IVD use.
Specifications
ColorColourless / Pale pink
DescriptionBrilliance CampyCount Agar
Fill Weight18mL
Format90mm Monoplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life8 Weeks
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2°C to 8°C