Product Image
Thermo Scientific™
Remel™ LB Agar w/Ampicillin (100μg/mL) (Luria-Bertani)
Use for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
R11020412mm x 85mm Monoplate
R11084615mm x 150mm Monoplate
Catalog number R110204
Price (USD)
-
Format:
12mm x 85mm Monoplate
Thermo Scientific™ Remel LB Agar with Ampicillin (100μg/mL) (Luria-Bertani) is used for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
DescriptionLB (Luria-Bertani) Agar w/Ampicillin 100micrograms/mL
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach