Product Image
Thermo Scientific™
Remel™ LB Agar w/Tetracycline, 25 μg/mL
Use for cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Have Questions?
Catalog NumberFormat
R110227Monoplate Deep Pour
Catalog number R110227
Price (USD)
-
Format:
Monoplate Deep Pour
Thermo Scientific™ Remel LB Agar with Tetracycline, 25μg/mL (Luria-Bertani) is for cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
DescriptionLB Agar w/Tetracycline, 25µg
FormatMonoplate Deep Pour
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach