Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
Thermo Scientific™
Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R111102100/Pk.
R11110120/Pk.
Catalog number R111102
Price (USD)
-
Quantity:
100/Pk.
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
  • Manufactured in a clean room environment to provide the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Not intended for IVD use.
Specifications
DescriptionContact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
FormatContact - Single Bag
Quantity100/Pk.
Product TypeAgar Plate
Unit SizeEach