Product Image
Thermo Scientific™
Remel™ Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6)
Thermo Scientific™ Remel Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6) is used for cultivation of yeast and fungi.
Have Questions?
Catalog NumberFormat
R111105Contact - Single Bag
Catalog number R111105
Price (USD)
-
Format:
Contact - Single Bag
Thermo Scientific™ Remel Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6) is used for cultivation of yeast and fungi.
  • Manufactured in a clean room environment to provide the highest level of sterility assurance
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
  • Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
Not intended for IVD use.
Specifications
DescriptionContact I Sabouraud Dextrose Agar
FormatContact - Single Bag
Quantity20/Pk.
Product TypeAgar Plate
Unit SizeEach