Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates
Thermo Scientific™

Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates

Obtain fast and accurate results with a contact plate designed for environmental monitoring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeIncludes
R11182010/Pk.55mm Contact PlateTryptic Soy AgarLecithin, Polysorbate 80
R111822100/Pk.55mm Contact PlateTryptic Soy AgarLecithin, Polysorbate 80
Catalog number R111820
Price (MXN)
-
Quantity:
10/Pk.
Format:
55mm Contact Plate
Media Type:
Tryptic Soy Agar
Includes:
Lecithin, Polysorbate 80
Effortlessly sample your environment using Thermo Scientific™ Isolator Wrap™ Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 Contact Plates.
Isolator Wrap plates are for environmental sampling. These products are manufactured in a cleanroom environment and validated sterile by gamma radiation. Contact plates use GripFit™ technology, which ensures the lid remains in place during handling. Plates are shrink-wrapped and contained in a protective outer barrier wrap, which is impermeable to isolator decontamination procedures but maintains the integrity of the medium inside. An extra sterile bag is provided for transporting the plates to the laboratory for testing.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
AdditiveLecithin, Polysorbate 80
Certifications/ComplianceISO 7/8 (Grade C and D)
ColorClear agar light to med amber
Country of OriginUnited States
Diameter (Metric)55 mm
Fill Volume15 mL
For Use With (Application)Environmental Monitoring
Format55mm Contact Plate
IncludesLecithin, Polysorbate 80
Media TypeTryptic Soy Agar
Packaging TypeFlow Wrapped - Single Layer and Extra Sterile Bag
Quantity10/Pk.
Shelf Life182 Days
FormPrepared Medium
Manufacturing OriginUnited States
Product TypeAgar Plate
SterilitySterile
Test TypeEnvironmental Test
Unit SizeEach
Contents & Storage
2°C to 8°C