Product Image
Thermo Scientific™
Remel™ Phosphate Buffer w/1% Peptone
Thermo Scientific™ Remel Phosphate Buffer w/1% Peptone is used as a pre-enrichment for isolation of Salmonella from various food sources.
Have Questions?
Catalog NumberQuantityFormatMedia Type
R11208810 x 90 mLPB - 90 mLBuffered Peptone Water
Catalog number R112088
Price (USD)
-
Quantity:
10 x 90 mL
Format:
PB - 90 mL
Media Type:
Buffered Peptone Water
Thermo Scientific™ Remel Phosphate Buffer w/1% Peptone (Buffered Peptone Water) is used as a pre-enrichment for isolation of Salmonella from various food sources.
Not intended for IVD use.
Specifications
FormatPB - 90 mL
Includes1% Peptone
Media TypeBuffered Peptone Water
Quantity10 x 90 mL
Product TypeBuffer Solution
Unit SizeEach