Double Bagged Bottled D/E (Dey-Engley) Neut Broth
Thermo Scientific™

Double Bagged Bottled D/E (Dey-Engley) Neut Broth

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.
Have Questions?
Catalog NumberFormat
R112170Double Bagged Borosilicate Bottle
Catalog number R112170
Price (USD)
-
Format:
Double Bagged Borosilicate Bottle
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled D/E Neut Broth for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled D/E Neut Broth is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Add Remel Double Bagged Bottled D/E Neut Broth to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.

Complete Solutions, Complete Confidence:

  • Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media
  • Regulatory compliant: USP, JP, and EU harmonized media  
  • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs
  • Quality guaranteed: Certificates of Analysis with every lot produced  
  • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle
  • Multi-purpose media: suitable for general quality testing
Customizable formats and media types are available. Please inquire.

Remel D/E Neutralizing Broth is a liquid medium recommended for use in qualitative procedures for environmental sampling when neutralization of a chemical disinfectant is important to distinguish bacteriostatic and bactericidal activity.

The medium was formulated by Engley and Dey in 1970 for the neutralization and testing of antiseptics and disinfectants1. It is recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling.2 The medium will neutralize a broad spectrum of antiseptic and disinfectant chemicals, including mercurial, iodine and chlorine preparations, quaternary ammonium compounds, phenolics, formaldehydes and glutaraldehydes.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use
For Laboratory Use

General References:

  1. Engley, F.B. and B.P. Dey 1970. Chemical Specialities Manufacturing Association Proceedings.
  2. Downes,F.P. and K.Ito.2001. Compendium of Methods for the Microbiological Examination of Foods. 4th ed. APHA, Washington, D.C.
Specifications
Cap TypePolypropylene plastic cap w/ Teflon
DescriptionDouble Bagged Bottled D/E Neut Broth
FormatDouble Bagged Borosilicate Bottle
Packaging Type90mL
Quantity10 Pack
Shelf Life182 Days
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 8°C